Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSENEN/PEN2 Antibody (JE41-28), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP275632
Description
PSENEN/PEN2 Monoclonal specifically detects PSENEN/PEN2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PSENEN/PEN2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200 | |
gamma-secretase subunit PEN-2, MSTP064, PEN-2, PEN2hematopoietic stem/progenitor cells protein MDS033, presenilin enhancer 2 homolog (C. elegans), Presenilin enhancer protein 2 | |
Rabbit | |
12 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
JE41-28 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
PSENEN | |
Recombinant protein within Human PSENEN/PEN2 aa 1-101/ 101. (SwissProt: Q9NZ42 Human; SwissProt: Q9CQR7 Mouse; SwissProt: O88777 Rat) | |
Protein A purified | |
Growth and Development, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
55851.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction