Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Aeruginosa Antibody (1001/214) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$181.90 - $487.50

Specifications

Antigen Pseudomonas Aeruginosa
Clone 1001/214
Concentration 1.0 mg/mL
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Applications Immunocytochemistry, ELISA, Immunofluorescence
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018467
SDP
View Documents
Novus Biologicals
NBP1259460.025MG
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP125946
SDP
View Documents
Novus Biologicals
NBP125946
0.1 mg
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Pseudomonas Aeruginosa
1.0 mg/mL
Immunocytochemistry, ELISA, Immunofluorescence
Unconjugated
Mouse
Bacteria
P. aeruginosa serotype 3 (ATCC 33350)
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1001/214
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Monoclonal
Purified
RUO
Pseudomonas aeruginosa 9
IgG3
Protein A purified
NBP1-25946 is specific for P. aeruginosa serotype 3.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.