Learn More
Pseudomonas Aeruginosa Antibody (1001/214), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP125946H
Catalog No. NBP125946H
Only null left
Add to Cart
Description
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA.
Specifications
Specifications
Pseudomonas Aeruginosa
Monoclonal
HRP
PBS with No Preservative
Mouse
Protein A purified
RUO
NBP1-25946 is specific for P. aeruginosa serotype 3.
Store at 4C in the dark.
IgG3
ELISA
1001/214
ELISA
Pseudomonas aeruginosa 9
P. aeruginosa serotype 3 (ATCC 33350)
0.1 mL
Primary
Bacteria
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.