Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pseudomonas Aeruginosa Antibody (1200/472), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308420JF646
Description
Pseudomonas Aeruginosa Monoclonal antibody specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISASpecifications
Pseudomonas Aeruginosa | |
Monoclonal | |
Janelia Fluor 646 | |
Pseudomonas aeruginosa 9 | |
Pseudomonas aeruginosa serotype 6C | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
1200/472 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction