Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pseudomonas Aeruginosa Antibody (95/159), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064562AF532
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Pseudomonas Aeruginosa | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Protein A purified | |
Primary | |
Bacteria | |
IgG1 |
ELISA, Immunocytochemistry, Immunofluorescence | |
95/159 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Pseudomonas aeruginosa 9 | |
Pseudomonas aeruginosa, serotype 9 (NCTC 11448, ATCC 33356). | |
0.1 mL | |
Specific for Pseudomonas aeruginosa, serotype 9. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction