Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pseudomonas Aeruginosa Antibody (95/159), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064562UV
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Pseudomonas Aeruginosa | |
Monoclonal | |
DyLight 350 | |
Pseudomonas aeruginosa 9 | |
Pseudomonas aeruginosa, serotype 9 (NCTC 11448, ATCC 33356). | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
95/159 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Specific for Pseudomonas aeruginosa, serotype 9. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction