Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Aeruginosa Antibody (B11), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. N104071A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N104071A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N104071A94 Supplier Novus Biologicals Supplier No. NB11040711AF594
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Pseudomonas Aeruginosa
Applications ELISA
Classification Monoclonal
Clone B11
Conjugate Alexa Fluor 594
Dilution ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Pseudomonas aeruginosa 9
Host Species Mouse
Immunogen Purified outer membrane protein of Pseudomonas aeruginosa.
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This Antibody recognises Pseudomonas aeruginosa a gram-negative, opportunistic pathogen, commonly found in soil and water.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.