Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pseudomonas Aeruginosa Antibody (B11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11040711H
Description
Pseudomonas Aeruginosa Monoclonal specifically detects Pseudomonas Aeruginosa in Bacteria samples. It is validated for ELISA.Specifications
Pseudomonas Aeruginosa | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein A purified | |
RUO | |
This Antibody recognises Pseudomonas aeruginosa a gram-negative, opportunistic pathogen, commonly found in soil and water. | |
Store at 4C in the dark. | |
IgG2a |
ELISA | |
B11 | |
ELISA | |
Pseudomonas aeruginosa 9 | |
Purified outer membrane protein of Pseudomonas aeruginosa. | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction