Learn More
Invitrogen™ Pseudomonas Polyclonal Antibody, Biotin

Description
The PA1-73119 antibody reacts with Pseudomonas species. PA1-73119 has been successfully used in immunofluorescence applications. Possible applications for this product include avidin and streptavidin amplification systems for immunohistochemistry, ELISA, fluorescence microscopy, and immunoblotting. The PA1-73119 antibody was raised against whole cells of Pseudomonas aeruginosa, ATCC #27853. Recommended short term storage of less than 6 months at 2-8ºC. For extended storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Pseudomonas |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Host Species | Rabbit |
| Immunogen | Whole cells of Pseudomonas aeruginosa, ATCC #27853. |
| Purification Method | Protein A |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.