Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pseudomonas sp. CPG2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117627
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117627 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117627 Supplier Invitrogen™ Supplier No. PA5117627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in direct-ELISA.

Catalyzes the hydrolysis of reduced and non-reduced folates to pteroates and L-glutamate. This enzyme has a broad specificity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pseudomonas sp. CPG2
Applications Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation Antibody with 0.1M NaCl and 0.015M sodium azide; pH 7.2
Gene Alias Carboxypeptidase G2; Folate hydrolase G2; Pteroylmonoglutamic acid hydrolase G2
Host Species Rabbit
Immunogen Recombinant Pseudomonas sp.Carboxypeptidase G2 protein (23-415aa).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.