Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pseudorabies Virus Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203534
Description
Pseudorabies Virus Polyclonal specifically detects Pseudorabies Virus in Human, Rat, Canine, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
| Pseudorabies Virus | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| Purified pseudorabies virus-Bartha. | |
| 100 μg | |
| Virology Bacteria and Parasites | |
| Pseudorabies Virus | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 1 mg/mL | |
| Western Blot 0.2 - 1.0 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Frozen 1.0 ug/ml | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction