Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP233825
Description
PSF2 Polyclonal specifically detects PSF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
PSF2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Q9Y248 | |
GINS2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: ELVTIIPNFSLDKIYLIGGDLGPFNPGLPVEVPLWLAINLKQRQKCRLLPPEWMDVEKLEKMRDHERKEETFTPMPSPYYMELTKLLLNHASDNIPKADEIRTLVKDMWDTRIAKLRVSADSFVRQQEAHAKLDNLTLMEINTSG | |
0.1 mL | |
Cell Cycle and Replication | |
51659 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DNA replication complex GINS protein PSF2, GINS complex subunit 2, GINS complex subunit 2 (Psf2 homolog), HSPC037, PSF2Pfs2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction