Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSF3 Antibody (OTI3A12), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273680C
Description
PSF3 Monoclonal specifically detects PSF3 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PSF3 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GINS3 | |
Full length recombinant protein of human PSF3 (NP_073607) produced in E.coli. | |
0.1 mL | |
64785.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI3A12 | |
Western Blot | |
DNA replication complex GINS protein PSF3, GINS complex subunit 3, GINS complex subunit 3 (Psf3 homolog), PSF3FLJ13912 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction