Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSG-1 Mouse anti-Human, Clone: 684701, R&D Systems™

Catalog No. MAB6799SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6799SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6799SP Supplier R&D Systems Supplier No. MAB6799SP
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PSG-1 Monoclonal antibody MAB6799SP specifically detects PSG-1 in Human samples. It is validated for Immunohistochemistry, Western Blotting

Specifications

Antigen Pan PSG
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 684701
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P11464
Gene Symbols PSG1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human PSG-1, Val36-Pro419, Accession # P11464
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5669
Test Specificity Detects human PSG-1, -2, -3, -5 and -6 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.