Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSG1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297038PE
Description
PSG1 Polyclonal antibody specifically detects PSG1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
PSG1 | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
B1G1CD66 antigen-like family member F, CD66f, CD66f antigen, DHFRP2, Fetal liver non-specific cross-reactive antigen 1/2, FLJ90598, FLJ90654, FL-NCA-1/2, PBG1, pregnancy specific beta-1-glycoprotein 1, pregnancy-specific B-1 glycoprotein, Pregnancy-specific beta-1 glycoprotein C/D, pregnancy-specific beta-1-glycoprotein 1, Pregnancy-specific glycoprotein 1, PS-beta-C/D, PS-beta-G-1, PSBG-1, PSBG1SP1, PSGGAPSG95, PSGIIA | |
Produced in rabbits immunized with purified, recombinant Human PSG1 (Accession#: NP_001171754.1; Met1-Pro419) | |
0.1 mL | |
CD Markers, Immunology | |
5669 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction