Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSG1 Monoclonal Antibody (5C6G7)
GREENER_CHOICE

Catalog No. PIMA538611
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538611 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538611 Supplier Invitrogen™ Supplier No. MA538611
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

Human pregnancy-specific glycoproteins (PSG), are a group of closely related secreted glycoproteins which are highly expressed in placental syncytiotrophoblast cells of fetal origin. PSG are commonly expressed in tumors of trophoblast origin (hydatidiform mole, choriocarcinoma). They represent the most abundant fetal protein in the maternal blood at term. The eleven human PSG genes (PSG1-PSG11) form together with the carcinoembryonic antigen (CEA)-related cell adhesion molecule (CEACAM) genes the CEACAM gene family. PSG stimulates secretion of TH2-type cytokines from monocytes. CD9 was shown to represent the monocyte receptor for murine PSG17. PSG are thought to modulate the maternal immune system during pregnancy thus protecting the semiallotypic fetus from rejection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 5C6G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PSG1
Gene Accession No. P11464
Gene Alias B1G1; CD66 antigen-like family member F; CD66f; DHFRP2; Fetal liver non-specific cross-reactive antigen 1/2; FLJ90598; FLJ90654; FL-NCA-1/2; PBG1; pregnancy specific beta-1-glycoprotein 1; pregnancy-specific B-1 glycoprotein; pregnancy-specific beta-1 glycoprotein C/D; Pregnancy-specific beta-1-glycoprotein 1; PS-beta-C/D; PS-beta-G-1; PSBG1; PSBG-1; PSG 1; PSG IIA-d; PSG1; PSG-1d; PSG95; PSGGA; PSGIIA; PSGIIA-a; PSGIIA-b; PSGIIA-c; SP 1; SP1
Gene Symbols PSG1
Host Species Mouse
Immunogen Purified recombinant fragment of human PSG1 (aa: 250-419) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5669
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.