Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSG11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 163521AP150UL

Description
PSG11, also named as PSG13, PSG14, PSBG-11 and PSBG-13, belongs to the immunoglobulin superfamily and CEA family. PSBG are produced in high quantity during pregcy. PSG11 and other PSGs induce dose-dependent secretion of anti-inflammatory cytokines by human monocytes.Specifications
PSG11 | |
Polyclonal | |
Unconjugated | |
PSG11 | |
PS beta G 11, PS beta G 13, PSBG 11, PSBG 13, PSG11, PSG13, PSG14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5680 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UQ72 | |
PSG11 | |
PSG11 Fusion Protein Ag9562 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction