Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Psg16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586002
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586002 Supplier Invitrogen™ Supplier No. PA586002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Psg16 Polyclonal Antibody for ICC/IF, Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen Psg16
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Psg16
Gene Accession No. 0
Gene Alias bCEA; carcinoembryonic antigen 11; Cea11; pregnancy specific glycoprotein 16; PSG; Psg16
Gene Symbols Psg16
Host Species Rabbit
Immunogen Synthetic peptide within mouse Psg16 aa 346-395.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26436
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.