Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSG4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549132
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549132 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549132 Supplier Invitrogen™ Supplier No. PA549132
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSG4 Polyclonal Antibody for Western Blot

The protein encoded by this gene is a pregnancy-specific glycoprotein (PSG), one of several encoded by a cluster of similar genes on chromosome 19. This gene is a member of the carcinoembryonic antigen (CEA) gene family and may play a role in regulation of the innate immune system. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PSG4
Gene Accession No. Q00888
Gene Alias CGM4; pregnancy specific beta-1-glycoprotein 4; pregnancy-specific beta-1-glycoprotein 4; Pregnancy-specific beta-1-glycoprotein 9; Pregnancy-specific beta-1-glycoprotein-4; pregnancy-specific glycoprotein 4; pregnancy-specific glycoprotein 9; PS-beta-G-4; PS-beta-G-9; PSBG-4; PSBG-9; PSG4; PSG9
Gene Symbols PSG4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 73-100 amino acids from the N-terminal region of human PSG4.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.