Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSG4 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501726698 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6698 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6698 Supplier Proteintech Group Inc Supplier No. 109022AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a pregcy-specific glycoprotein (PSG), one of several encoded by a cluster of similar genes on chromosome 19. This gene is a member of the carcinoembryonic antigen (CEA) gene family and may play a role in regulation of the innate immune system. Several transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen PSG4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PSG4
Gene Accession No. Q00888
Gene Alias CGM4, CGM4, PSG9, PS beta G 4, PS beta G 9, PSBG 4, PSBG 9, PSG4, PSG9
Gene Symbols PSG4
Host Species Rabbit
Immunogen PSG4 Fusion Protein Ag1167
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5672
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.