Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSG5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542096 Supplier Invitrogen™ Supplier No. PA542096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QLMDLYHYIT SYVVDGQINI YGPAYTGRET VYSNASLLIQ NVTREDAGSY Sequence homology: Dog: 77%; Horse: 77%; Human: 100%; Mouse: 85%; Rat: 85%.

The pregnancy-specific beta 1 glycoprotein (PSG) is a group of heterogeneous proteins produced in large amounts by the human syncytiotrophoblast. They belong to the carcinoembryonic antigen (CEA) family. The function of PSG5 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PSG5
Gene Accession No. Q15238
Gene Alias Fetal liver non-specific cross-reactive antigen 3; FL-NCA-3; pregnancy specific beta-1-glycoprotein 5; pregnancy-specific beta 1 glycoprotein; pregnancy-specific beta-1 glycoprotein; pregnancy-specific beta-1-glycoprotein 5; Pregnancy-specific beta-1-glycoprotein-5; pregnancy-specific glycoprotein 5; PS-beta-G-5; PSBG-5; PSG; PSG5
Gene Symbols PSG5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human PSG5 (aa 71-120).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5673
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.