Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSG6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598094
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598094 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598094 Supplier Invitrogen™ Supplier No. PA598094
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSG6 Polyclonal Antibody for Western Blot, ELISA

The human pregnancy-specific glycoproteins (PSGs) are a group of molecules that are mainly produced by the placental syncytiotrophoblasts during pregnancy. PSGs comprise a subgroup of the carcinoembryonic antigen (CEA) family, which belongs to the immunoglobulin superfamily. For additional general information about the PSG gene family, see PSG1 (MIM 176390).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PSG6
Gene Accession No. Q00889
Gene Alias CGM3; pregnancy specific beta-1-glycoprotein 6; pregnancy-specific beta-1-glycoprotein 10; Pregnancy-specific beta-1-glycoprotein 12; pregnancy-specific beta-1-glycoprotein 6; PS-beta-G-10; PS-beta-G-12; PS-beta-G-6; PSBG-10; PSBG-12; PSBG-6; PSG10; PSG12; PSG6; PSGGB
Gene Symbols PSG6
Host Species Rabbit
Immunogen Recombinant Human Pregnancy-specific beta-1-glycoprotein 6 protein (35-300AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5675
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.