Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524864
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524864 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524864 Supplier Invitrogen™ Supplier No. PA524864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSG7 Polyclonal Antibody for Western Blot, IHC (P), Flow

PSG7 is a member of the pregnancy-specific glycoprotein (PSG) gene family. The PSG genes are a subgroup of the carcinoembryonic antigen (CEA) family of immunoglobulin-like genes, and are found in a gene cluster at 19q13.1-q13.2 telomeric to another cluster of CEA-related genes. The PSG genes are expressed by placental trophoblasts and released into the maternal circulation during pregnancy, and are thought to be essential for maintenance of normal pregnancy. The reference genome contains a nonsense mutation that disrupts the coding sequence, suggesting that this gene may be evolving into a pseudogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSG7
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PSG7
Gene Accession No. Q13046
Gene Alias pregnancy specific beta-1-glycoprotein 7 (gene/pseudogene); pregnancy-specific glycoprotein 7; PS-beta-G-7; PSBG-7; PSG1; PSG7; PSGGA; putative pregnancy-specific beta-1-glycoprotein 7
Gene Symbols PSG7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 265-293 amino acids from the C-terminal region of human PSG7.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5676
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.