Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSG9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531707
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The human pregnancy-specific glycoproteins (PSGs) are a group of molecules that are mainly produced by the placental syncytiotrophoblasts during pregnancy. PSGs comprise a subgroup of the carcinoembryonic antigen (CEA) family, which belongs to the immunoglobulin superfamily. For additional general information about the PSG gene family, see PSG1 (MIM 176390).
Specifications
PSG9 | |
Polyclonal | |
Unconjugated | |
PSG9 | |
pregnancy specific beta-1-glycoprotein 11; pregnancy specific beta-1-glycoprotein 9; pregnancy-specific beta-1 glycoprotein B; Pregnancy-specific beta-1-glycoprotein 11; pregnancy-specific beta-1-glycoprotein 11 C-R domain; pregnancy-specific beta-1-glycoprotein 9; Pregnancy-specific glycoprotein 7; PS34; PS-beta-B; PS-beta-G-11; PS-beta-G-9; PSBG-11; PSBG-9; PSG11; PSG7; PSG9; PSGII | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5678 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.65 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q00887 | |
PSG9 | |
Recombinant protein fragment corresponding to a region within amino acids 220 and 402 of Human PSG9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction