Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSG9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107227
Description
PSG9 Polyclonal Antibody for Western Blot
The human pregnancy-specific glycoproteins (PSGs) are a group of molecules that are mainly produced by the placental syncytiotrophoblasts during pregnancy. PSGs comprise a subgroup of the carcinoembryonic antigen (CEA) family, which belongs to the immunoglobulin superfamily. For additional general information about the PSG gene family, see PSG1 (MIM 176390).
Specifications
| PSG9 | |
| Polyclonal | |
| Unconjugated | |
| PSG9 | |
| pregnancy specific beta-1-glycoprotein 11; pregnancy specific beta-1-glycoprotein 9; pregnancy-specific beta-1 glycoprotein B; Pregnancy-specific beta-1-glycoprotein 11; pregnancy-specific beta-1-glycoprotein 11 C-R domain; pregnancy-specific beta-1-glycoprotein 9; Pregnancy-specific glycoprotein 7; PS34; PS-beta-B; PS-beta-G-11; PS-beta-G-9; PSBG-11; PSBG-9; PSG11; PSG7; PSG9; PSGII | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 5678 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q00887 | |
| PSG9 | |
| Synthetic peptide directed towards the N terminal region of human PSG9. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction