Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSGL-1/CD162 Antibody (001), DyLight 680

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290714FR
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Mouse samples. It is validated for Flow Cytometry.Specifications
PSGL-1/CD162 | |
Monoclonal | |
DyLight 680 | |
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PSGL-1/CD162 (Q62170; Met1-Cys307). | |
0.1 mL | |
Cholesterol Metabolism, Immunology, Lipid and Metabolism | |
6404 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry | |
1 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction