Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSGL-1/CD162 Antibody (PSGL1/1601), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253344200UG
Description
PSGL-1/CD162 Monoclonal antibody specifically detects PSGL-1/CD162 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PSGL-1/CD162 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA | |
| Mouse | |
| 82 kDa | |
| 200 μg | |
| Cholesterol Metabolism, Immunology, Lipid and Metabolism | |
| 6404 | |
| Store at 4C. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PSGL1/1601 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| SELPLG | |
| Recombinant human full-length SELPLG (CD162) protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction