Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PSGL-1/CD162 Antibody, R&D Systems™
Mouse Monoclonal Antibody
$172.84 - $451.24
Specifications
Antigen | PSGL-1/CD162 |
---|---|
Clone | 688102 |
Dilution | Immunohistochemistry 5-25 ug/mL, Neutralization |
Applications | Immunohistochemistry, Neutralization |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3345SP
|
R&D Systems
MAB3345SP |
25 μg |
Each for $172.84
|
|
|||||
MAB3345100
|
R&D Systems
MAB3345100 |
100 μg |
Each for $451.24
|
|
|||||
Description
PSGL-1/CD162 Monoclonal specifically detects PSGL-1/CD162 in Human samples. It is validated for Immunohistochemistry, Neutralization.Specifications
PSGL-1/CD162 | |
Immunohistochemistry 5-25 ug/mL, Neutralization | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
CD162, CD162 antigen, P-selectin glycoprotein ligand 1, PSGL1, PSGL-1cutaneous lymphocyte-associated associated antigen, selectin P ligandCLA, SELPLG | |
Chinese hamster ovary cell line CHO-derived recombinant human PSGL-1, Gln42-Gly295, Accession # Q14242 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human PSGL-1 in direct ELISAs. |
688102 | |
Immunohistochemistry, Neutralization | |
Unconjugated | |
Mouse | |
Human | |
Q14242 | |
6404 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title