Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSGR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575357 Supplier Invitrogen™ Supplier No. PA575357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

PSGR (prostate-specific G-protein coupled receptor), also known as OR51E2 (olfactory receptor 51E2), is a 320 amino acid multi-pass membrane protein that belongs to the olfactory receptor subfamily of G-protein coupled receptors. Expressed exclusively in prostate tissue and upregulated in prostate cancer, PSGR functions as an odorant receptor that binds odorant molecules and triggers the perception of smell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSGR
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene OR51E2
Gene Accession No. O88628, Q8VBV9, Q9H255
Gene Alias 4633402A21Rik; GA_x6K02T2PBJ9-5459657-5458695; G-protein coupled receptor RA1c; hCG_2044604; HPRAJ; Mol2.3; MOR18-2; olfactory receptor 51E2; Olfactory receptor 59; olfactory receptor 78; olfactory receptor family 51 subfamily E member 2; olfactory receptor MOR18-2; olfactory receptor OR11-16; olfactory receptor, family 51, subfamily E, member 2; olfactory receptor, family 51, subfamily E, member 3 pseudogene; olfactory receptor, family 52, subfamily A, member 2; Olfr78; Olr59; OR51E2; OR51E3P; OR52A2; prostate specific G-protein coupled receptor; prostate-specific G-protein coupled receptor; PSGR; RA1c
Gene Symbols Olfr78, Olr59, OR51E2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 202-250 of Human PSGR.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170639, 170816, 81285
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.