Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554060
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554060 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554060 Supplier Invitrogen™ Supplier No. PA554060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KQVETEEAGV VTTATASVNL KVSPKRGRPA ATEVKIPKPR GRPKMVKQPC PSESDIITEE DKSKKKGQEE KQPKKQPKKD EEGQ Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.

Transcriptional coactivator involved in neuroepithelial stem cell differentiation and neurogenesis. Involved in particular in lens epithelial cell gene regulation and stress responses. May play an important role in lens epithelial to fiber cell terminal differentiation. May play a protective role during stress-induced apoptosis. Isoform 2 is a more general and stronger transcriptional coactivator. Isoform 2 may also act as an adapter to coordinate pre-mRNA splicing. Cellular cofactor for lentiviral integration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSIP1
Applications ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSIP1
Gene Accession No. O75475
Gene Alias AA408851; AU015605; CLL-associated antigen KW-7; dense fine speckles 70; dense fine speckles 70 kDa protein; DFS 70; DFS70; LEDGF; Ledgfa; Ledgfb; Lens epithelium-derived growth factor; mLEDGF; p52; p75; PAIP; PC4 and SFRS1 interacting protein 1; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; PSIP1; Psip2; SBP75; supercoiled DNA binding protein 75; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52
Gene Symbols PSIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSIP1. Recombinant protein control fragment (Product #RP-92573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11168
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.