Learn More
Invitrogen™ PSIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117136
Description
Positive Samples: HeLa, HepG2, Jurkat, SW480, NIH/3T3, Mouse brain, Mouse thymus Immunogen sequence: NMLKGQHEKE AADRKRKQEE QMETEQQNKD EGKKPEVKKV EKKRETSMDS RLQRIHAEIK NSLKIDNLDV NRCIEALDEL ASLQVTMQQA QKHTEMITTL KKIRRFKVSQ VIMEKSTMLY NKFKNMFLVG EGDSVITQVL NKSLAEQRQH EEANKTKDQG KKGPNKKLEK EQTGSKTLNG GSDAQDGNQP QHNGESNEDS KDNHEASTKK KPSSEERETE ISLKDSTLDN.
Transcriptional coactivator involved in neuroepithelial stem cell differentiation and neurogenesis. Involved in particular in lens epithelial cell gene regulation and stress responses. May play an important role in lens epithelial to fiber cell terminal differentiation. May play a protective role during stress-induced apoptosis. Isoform 2 is a more general and stronger transcriptional coactivator. Isoform 2 may also act as an adapter to coordinate pre-mRNA splicing. Cellular cofactor for lentiviral integration.
Specifications
| PSIP1 | |
| Polyclonal | |
| Unconjugated | |
| PSIP1 | |
| AA408851; AU015605; CLL-associated antigen KW-7; dense fine speckles 70; dense fine speckles 70 kDa protein; DFS 70; DFS70; LEDGF; Ledgfa; Ledgfb; Lens epithelium-derived growth factor; mLEDGF; p52; p75; PAIP; PC4 and SFRS1 interacting protein 1; PC4 and SFRS1 interacting protein 2; PC4 and SFRS1-interacting protein; PSIP1; Psip2; SBP75; supercoiled DNA binding protein 75; transcriptional coactivator p52/p75; Transcriptional coactivator p75/p52 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 101739, 11168, 313323 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75475, Q812D1, Q99JF8 | |
| PSIP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 301-530 of human PSIP1 (NP_1500912). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.