Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSKH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553278 Supplier Invitrogen™ Supplier No. PA553278
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SGFLPFDDES QTRLYRKILK GKYNYTGEPW PSISHLAKDF IDKLLILEAG HRMSAGQALD HPWVITMAAG SSMKNLQRAI SRNLMQRASP HSQSPGSAQS SKSHYSHKSR Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.

The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine (Ser/Thr) protein kinases. PSKH2 (protein serine kinase H2) is a 385 amino acid protein belonging to the CAMK Ser/Thr protein kinase family. Containing one protein kinase domain, PSKH2 is encoded by a gene located on human chromosome 8, which consists of nearly 146 million base pairs, houses more than 800 genes and is associated with a variety of diseases and malignancies. Schizophrenia, bipolar disorder, Trisomy 8, Pfeiffer syndrome, congenital hypothyroidism, Waardenburg syndrome and some leukemias and lymphomas are thought to occur as a result of defects in specific genes that map to chromosome 8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSKH2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PSKH2
Gene Accession No. Q96QS6
Gene Alias protein serine kinase H2; PSKH2; PSK-H2; serine/threonine kinase PSKH2; serine/threonine-protein kinase H2
Gene Symbols PSKH2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PSKH2. Recombinant protein control fragment (Product #RP-91450).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85481
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.