Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA/FOLH1/NAALADase I Antibody (k1H7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104338
Description
PSMA/FOLH1/NAALADase I Monoclonal specifically detects PSMA/FOLH1/NAALADase I in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
PSMA/FOLH1/NAALADase I | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000-1:3000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
Cell growth-inhibiting gene 27 protein, cell growth-inhibiting protein 27, EC 3.4.17.21, folate hydrolase (prostate-specific membrane antigen) 1, Folate hydrolase 1, FOLHNAALADase I, Folylpoly-gamma-glutamate carboxypeptidase, GCP2, GCPIImGCP, glutamate carboxylase II, glutamate carboxypeptidase 2, Glutamate carboxypeptidase II, Membrane glutamate carboxypeptidase, NAALAD1N-acetylated-alpha-linked acidic dipeptidase I, NAALAdase, N-acetylated alpha-linked acidic dipeptidase 1, prostate specific membrane antigen variant F, Prostate-specific membrane antigen, PSMAFGCP, PSMPteroylpoly-gamma-glutamate carboxypeptidase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human PSMA mAb, clone k1H7, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PSMA protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
k1H7 | |
Unconjugated | |
Q04609 | |
FOLH1 | |
Recombinant human PSMA (117-351aa) purified from E. coli (NP_001014986). | |
0.1 mL | |
Cancer, Prostate Cancer | |
2346 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction