Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMA Recombinant Rabbit Monoclonal Antibody (SP29)
GREENER_CHOICE

Catalog No. PIMA516395
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516395 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516395 Supplier Invitrogen™ Supplier No. MA516395
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is prostate carcinoma.

This gene encodes a type II transmembrane glycoprotein belonging to the M28 peptidase family. The protein acts as a glutamate carboxypeptidase on different alternative substrates, including the nutrient folate and the neuropeptide N-acetyl-l-aspartyl-l-glutamate and is expressed in a number of tissues such as prostate, central and peripheral nervous system and kidney. A mutation in this gene may be associated with impaired intestinal absorption of dietary folates, resulting in low blood folate levels and consequent hyperhomocysteinemia. Expression of this protein in the brain may be involved in a number of pathological conditions associated with glutamate excitotoxicity. In the prostate the protein is up-regulated in cancerous cells and is used as an effective diagnostic and prognostic indicator of prostate cancer. This gene likely arose from a duplication event of a nearby chromosomal region. Alternative splicing gives rise to multiple transcript variants encoding several different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMA
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone SP29
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene FOLH1
Gene Accession No. Q04609
Gene Alias Cell growth-inhibiting gene 27 protein; FGCP; folate hydrolase (prostate-specific membrane antigen) 1; folate hydrolase 1; FOLH; Folh1; Folylpoly-gamma-glutamate carboxypeptidase; GCP2; GCPII; GIG27; glutamate carboxylase II; glutamate carboxypeptidase 2; Glutamate carboxypeptidase II; membrane glutamate carboxypeptidase; mGCP; mopsm; Naalad; NAALAD1; NAALAdase; NAALADase I; N-acetylated alpha-linked acidic dipeptidase; N-acetylated alpha-linked acidic dipeptidase 1; N-acetylated-alpha-linked acidic dipeptidase I; prostate specific membrane antigen; prostate specific membrane antigen variant F; prostate-specific membrane antigen; Prostate-specific membrane antigen homolog; PSM; PSMA; pteroylpoly-gamma-glutamate carboxypeptidase
Gene Symbols FOLH1
Host Species Rabbit
Immunogen Synthetic peptide derived from C-terminus of human PSMA.
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2346
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.