Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA2 Antibody (OTI3D9), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273687AF594
Description
PSMA2 Monoclonal specifically detects PSMA2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PSMA2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
5683.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3D9 | |
Western Blot, Immunohistochemistry | |
PSMA2 | |
Recombinant protein fragment corresponding to amino acids 50-234 of human PSMA2 (NP_002778) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction