Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA2 Antibody (OTI3D9) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273687
Description
PSMA2 Monoclonal specifically detects PSMA2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Proteasome 20S alpha 2 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:500 | |
EC 3.4.25.1, HC3PSC2, Macropain subunit C3, MU, Multicatalytic endopeptidase complex subunit C3, PMSA2, proteasome (prosome, macropain) subunit, alpha type, 2, Proteasome component C3, proteasome subunit alpha type-2, proteasome subunit HC3, PSC3 | |
Mouse | |
25.7 kDa | |
100 μg | |
5683 | |
Human, Mouse, Rat, Canine, Monkey | |
Lyophilized |
Western Blot, Immunohistochemistry | |
OTI3D9 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PSMA2 | |
Human recombinant protein fragment corresponding to amino acids 50-234 of human PSMA2(NP_002778) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction