Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA2 Antibody (SR1404), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32209450UL
Description
PSMA2 Monoclonal antibody specifically detects PSMA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
PSMA2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
SR1404 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
HC3;MU;PMSA2;Proteasome subunit alpha type;Proteasome subunit alpha type-2;PSC2 | |
A synthesized peptide derived from human PSMA2 (Uniprot #: P25787) | |
50 μg | |
Stem Cell Markers | |
5683 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction