Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA2 Monoclonal Antibody (OTI3E2), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA505475S


Description
PSMA2 Monoclonal antibody specifically detects PSMA2 in Human samples. It is validated for Western BlotSpecifications
PSMA2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P25787 | |
Mouse | |
Affinity chromatography | |
RUO | |
5683 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI3E2 | |
Unconjugated | |
Psma2 | |
HC3; Lmpc3; Macropain subunit C3; MU; multicatalytic endopeptidase complex subunit C3; OTTHUMP00000207364; OTTHUMP00000207366; PMSA2; proteasome (prosome, macropain) subunit, alpha type 2; proteasome (prosome, macropain) subunit, alpha type, 2; proteasome 20S subunit alpha 2; proteasome alpha 2 subunit; proteasome component C3; proteasome subunit alpha 2; proteasome subunit alpha type, 2; proteasome subunit alpha type-2; proteasome subunit HC3; PSC2; PSC3; PSMA 2; PSMA2; wu:faa49c06; wu:fb98h03; zgc:110710 | |
Human recombinant protein fragment corresponding to amino acids 50-234 of human PSMA2 produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction