Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA2 Rabbit anti-Human, Mouse, Rat, Clone: 6L5H4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31677820UL
This item is not returnable.
View return policy
Description
PSMA2 Monoclonal antibody specifically detects PSMA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| PSMA2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human PSMA2 (P25787). | |
| 20 μg | |
| Stem Cell Markers | |
| 5683 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 6L5H4 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction