Learn More
Invitrogen™ PSMA3 Polyclonal Antibody

Description
Immunogen sequence: FRSNFGYNIP LKHLADRVAM YVHAYTLYSA VRPFGCSFML GSYSVNDGAQ LYMIDPSGVS YGYWGCAIGK ARQAAKTEIE KLQMKEMTCR DIVKEVAKII YIVHDEVKDK AFELELSWVG ELTNGRHEIV PKDIREEAEK YAKESL Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.
Specifications
Specifications
| Antigen | PSMA3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | Psma3 |
| Gene Accession No. | O70435, P18422, P25788 |
| Gene Alias | HC8; Lmpc8; Macropain subunit C8; multicatalytic endopeptidase complex subunit C8; multicatalytic proteinase subunit K; proteasome (prosome, macropain) subunit, alpha type 3; proteasome (prosome, macropain) subunit, alpha type, 3; proteasome component C8; proteasome subunit alpha 3; proteasome subunit alpha type-3; proteasome subunit C8; proteasome subunit K; PSA3; PSC3; PSC8; PSMA3; Psma3l; testicular secretory protein Li 43 |
| Gene Symbols | Psma3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.