Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMA3 Recombinant Rabbit Monoclonal Antibody (JE51-59)
GREENER_CHOICE

Catalog No. PIMA534879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534879 Supplier Invitrogen™ Supplier No. MA534879
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: A549, PC-12, NIH/3T3, SW620, rat large intestine tissue, human liver tissue, human breast tissue, human placenta tissue.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. Two alternative transcripts encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMA3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE51-59
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene Psma3
Gene Accession No. P25788
Gene Alias HC8; Lmpc8; Macropain subunit C8; multicatalytic endopeptidase complex subunit C8; multicatalytic proteinase subunit K; proteasome (prosome, macropain) subunit, alpha type 3; proteasome (prosome, macropain) subunit, alpha type, 3; proteasome component C8; proteasome subunit alpha 3; proteasome subunit alpha type-3; proteasome subunit C8; proteasome subunit K; PSA3; PSC3; PSC8; PSMA3; Psma3l; testicular secretory protein Li 43
Gene Symbols Psma3
Host Species Rabbit
Immunogen Recombinant protein within Human PSMA3 aa 1-189.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5684
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.