Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMA4 Antibody (OTI4B8), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273690G
Description
PSMA4 Monoclonal specifically detects PSMA4 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PSMA4 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PSMA4 | |
Full length recombinant protein of human PSMA4 (NP_002780) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4B8 | |
Western Blot, Immunohistochemistry | |
HC9EC 3.4.25.1, HsT17706, Macropain subunit C9, MGC12467, MGC24813, Multicatalytic endopeptidase complex subunit C9, proteasome (prosome, macropain) subunit, alpha type, 4, Proteasome component C9, proteasome subunit alpha type-4, proteasome subunit HC9, Proteasome subunit L, PSC9MGC111191 | |
Mouse | |
Affinity Purified | |
Proteases & Other Enzymes | |
5685.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction