Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA4 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene
SDP

Catalog No. 502540553
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0553 30 μL
50-167-4796 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0553 Supplier OriGene Supplier No. TA504023S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMA4 Monoclonal antibody specifically detects PSMA4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PSMA4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1H10
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NABP2
Gene Accession No. Q9BQ15
Gene Alias C9; HC9; HsT17706; LOW QUALITY PROTEIN: proteasome subunit alpha type-4; macropain subunit C9; MGC111191; MGC12467; MGC24813; Multicatalytic endopeptidase complex subunit C9; multicatalytic proteinase subunit L; proteasome (prosome, macropain) subunit, alpha type 4; proteasome (prosome, macropain) subunit, alpha type, 4; proteasome 20S subunit alpha 4; proteasome alpha 4 subunit; proteasome component C9; proteasome subunit alpha 4; proteasome subunit alpha type-4; proteasome subunit alpha type-4-like protein; proteasome subunit HC9; proteasome subunit L; PSC9; PSMA 4; PSMA4; QtsA-10816; unnamed protein product; wu:fe05d10; zgc:56176
Host Species Mouse
Immunogen Full length human recombinant protein of human OBFC2B produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79035
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.