Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585394 Supplier Invitrogen™ Supplier No. PA585394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (98%), Xenopus laevis (98%), Pig (100%), Chicken (99%), Rhesus Monkey (100%), Bovine (100%). Positive Control: 293T, A431, HeLa, HepG2, PC-12, Rat-2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMA5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene PSMA5
Gene Accession No. P28066, P34064, Q9Z2U1
Gene Alias epididymis tissue sperm binding protein Li 11n; macropain subunit zeta; macropain zeta chain; Multicatalytic endopeptidase complex zeta chain; proteasome (prosome, macropain) subunit, alpha type 5; proteasome (prosome, macropain) subunit, alpha type, 5; proteasome alpha 5 subunit; proteasome component 5; proteasome subunit alpha 5; proteasome subunit alpha type-5; proteasome subunit alpha type-5-like protein; proteasome subunit zeta; proteasome zeta chain; PSC5; Psma5; ZETA
Gene Symbols PSMA5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PSMA5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26442, 29672, 5686
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.