Learn More
PSMA6 Mouse anti-Human, Clone: OTI1C4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF800108

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. A pseudogene has been identified on the Y chromosome.Specifications
PSMA6 | |
Monoclonal | |
Unconjugated | |
PSMA6 | |
IOTA, PROS27, p27K | |
Mouse | |
Affinity Chromatography | |
RUO | |
5687 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P60900 | |
PSMA6 | |
Full length human recombit protein of human PSMA6 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.