Learn More
PSMA6 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | PSMA6 |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-6117
![]() |
Bethyl Laboratories, Inc
A303809AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predict
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit. A pseudogene has been identified on the Y chromosome.Specifications
PSMA6 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
27 kDa prosomal protein; PROS-27; macropain iota chain; macropain subunit iota; multicatalytic endopeptidase complex iota chain; prosomal P27K protein; proteasome iota chain; proteasome subunit alpha type-6; proteasome subunit iota | |
Between 196 and 246 | |
Primary | |
4° C | |
PSMA6 |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P60900, Q9QUM9 | |
26443, 5687 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.