Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA8 Monoclonal Antibody (2G2G3), Proteintech
SDP

Catalog No. 502334199 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-233-4199 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-233-4199 Supplier Proteintech Group Inc Supplier No. 681231IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PSMA8 Monoclonal antibody specifically detects PSMA8 in Human, Mouse, Pig, Rabbit, Rat samples. It is validated for Western Blot

Specifications

Antigen PSMA8
Applications Western Blot
Classification Monoclonal
Clone 2G2G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Accession No. Q8TAA3
Gene Alias 2410072D24Rik; PSMA7L; PSMA8; RGD1311659
Host Species Mouse
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100338566
Target Species Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.