Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMA8, Mouse anti-Human, Clone: 2E3, Abnova™

Catalog No. 50859905 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-859-905 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-859-905 Supplier Abnova Corporation Supplier No. H00143471M03

Mouse Monoclonal Antibody

Sequence: GFDDDGISRLYQTDPSGTYHAWKANAIGRSAKTVREFLEKNYTEDAIASDSEAIKLAIKALLEVVQSGGRNIELAIIRRNQPLKMFSAKEVELYVTEIEK

Specifications

Antigen PSMA8
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene proteasome (prosome, macropain) subunit, alpha type, 8
Gene Accession No. NM_144662
Gene Alias MGC26605/PSMA7L
Gene Symbols PSMA8
Host Species Mouse
Immunogen PSMA8 (NP_653263, 143 a.a. ∼ 242 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143471
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.