Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PSMA8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589783
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589783 Supplier Invitrogen™ Supplier No. PA589783
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASRYDRAIT VFSPDGHLFQ VEYAQEAVKK GSTAVGIRGT NIVVLGVEKK SVAKLQDERT VRKICALDDH VCMAFAVLTI FIGLTADARV VINRARVECQ SHKLTVEDPV TVEYITRFIA TLKQKYTQSN GRRPFGISAL IVGFDDDGIS RLYQTDPSGT YHAWKANAIG RSAKTVREFL EKNYTEDAIA SDSEAIKLAI KALLEVVQSG GKNIELAIIR RNQPLKMFSA KEVELYVTEI; Positive Samples: K-562, Mouse thymus, Mouse testis, Rat testis; Cellular Location: Cytoplasm, Nucleus.

Component of the spermatoproteasome, a proteasome specifically found in testis that promotes acetylation-dependent degradation of histones, thereby participating actively to the exchange of histones during spermatogenesis. The proteasome is a protein complexe that degrades unneeded or damaged proteins by proteolysis, a chemical reaction that breaks peptide bonds. Required for 20S core proteasome assembly, essential for the degradation of meiotic proteins RAD51 and RPA1 at late prophase I and the progression of meiosis I during spermatogenesis. Localizes to the synaptonemal complex, a 'zipper'-like structure that holds homologous chromosome pairs in synapsis during meiotic prophase I. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMA8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PSMA8
Gene Accession No. Q8TAA3, Q9CWH6
Gene Alias 2410072D24Rik; proteasome (prosome, macropain) subunit, alpha type, 8; proteasome 20S subunit alpha 8; proteasome alpha 8 subunit; proteasome subunit alpha 8; proteasome subunit alpha type 7; proteasome subunit alpha type 7-like protein variant 1; proteasome subunit alpha type-7-like; Proteasome subunit alpha-type 7-like; Proteasome subunit alpha-type 8; psma7; PSMA7L; PSMA8; RGD1311659; wu:fk28c01; zgc:77139
Gene Symbols PSMA8
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human PSMA8 (NP_6532632).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143471, 364814, 73677
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.