Learn More
Invitrogen™ PSMAL Polyclonal Antibody

Description
PSMAL Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | PSMAL |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FOLH1B |
| Gene Accession No. | Q9HBA9 |
| Gene Alias | cell growth-inhibiting gene 26 protein; folate hydrolase 1B; folate hydrolase 2; folate hydrolase pseudogene; FOLH1B; FOLH2; FOLHP; GCP3; GCPIII; GIG26; glutamate carboxypeptidase III; NAALADase; N-acetylated-alpha-linked-acidic dipeptidase; Prostate-specific membrane antigen-like protein; PSM; PSMAL; PSMA-LIKE; Putative folate hydrolase 1B; putative N-acetylated-alpha-linked acidic dipeptidase |
| Gene Symbols | FOLH1B |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.